Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ AVPR1a-NFAT-bla CHO-K1 Cells

Catalog No. K1715
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 106 cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1715 2 x 106 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1715 Supplier Thermo Scientific™ Supplier No. K1715
Only null left
Add to Cart
Add to Cart
Request a Quote

GeneBLAzer™ AVPR1a-NFAT-bla CHO-K1 cells contain the human Arginine Vasopressin Receptor 1a (AVPR1a) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

GeneBLAzer™ AVPR1a-NFAT-bla CHO-K1 cells contain the human Arginine Vasopressin Receptor 1a (AVPR1a) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.

The GeneBLAzer™ AVPR1a-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Vasopressin. In addition, GeneBLAzer™ AVPR1a-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

The family of receptors that recognize vasopressin and oxytocin consists of four members: vasopressin receptors 1a (AVPR1a), 1b (AVPR1b), and 2 (AVPR2), and the oxytocin (OT) receptor. AVPR1a, AVPR1b, and the OT receptor couple to the Gq/G11 class of G proteins and trigger the release of intracellular calcium when stimulated. AVPR2 receptors couple to Gs proteins and activate adenyl cyclase. All three AVPR receptors have high affinity for their endogenous ligand, vasopressin, but AVPR1b has less affinity for oxytocin than does AVPR1a or AVPR2. The hormones vasopressin and oxytocin are synthesized in the hypothalamus. Oxytocin causes contractions in the uterus during childbirth. The main functions of arginine vasopressin (AVP) are reabsorption of water by the kidneys and contraction of smooth muscle in arteries. This system may also play a role in memory and learning.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000706.3
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ AVPR1a-NFAT-bla CHO-K1 cells
• GeneBLAzer™ AVPR1a-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ AVPR1a-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol AVPR1a
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry AVPR1A
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.