Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ CALCR -CRE-bla FreeStyle™ 293F Cells

Catalog No. K1480
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 106 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1480 2 x 106 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1480 Supplier Thermo Scientific™ Supplier No. K1480

Please call customer service for current pricing. 1-800-766-7000

The GeneBLAzer™ CALCR-CRE-bla Freestyle293 cells contain the CALCR receptor isoform (CTR2/CT(a)/CTRI1-), Accession No. NM_001742.2, stably integrated into the CellSensor™ CRE-bla Freestyle293 cell line.

The GeneBLAzer™ CALCR-CRE-bla Freestyle293 cells contain the CALCR receptor isoform (CTR2/CT(a)/CTRI1-), Accession No. NM_001742.2, stably integrated into the CellSensor™ CRE-bla Freestyle293 cell line. The CellSensor™ CRE-bla Freestyle293 cells contain a beta-lactamase (bla) reporter gene under control of the cAMP (CRE) response element.

CALCR-CRE-bla Freestyle293 cells have been tested for variable assay conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time and functionally validated for Z' and EC50 concentrations of calcitonin. Additional testing data using alternate stimuli are also provided. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_001742.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ CALCR-CRE-bla Freestyle™ 293F cells
• GeneBLAzer™ CALCR-CRE-bla Freestyle™ 293F Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ CALCR-CRE-bla Freestyle™ 293F cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
Gene Symbol CALCR2
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry CALCR2
Cell Line FreeStyle 293-F Cells
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.