Learn More
Thermo Scientific™ GeneBLAzer™ D1-CRE-bla CHO-K1 Cells
Description
GeneBLAzer™ D1-CRE-bla CHO-K1 cells contain the human Dopamine Receptor 1 (D1) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.
The GeneBLAzer™ D1-CRE-bla CHO-K1cells are functionally validated for Z' and EC50 concentrations of dihydrexidine. In addition, GeneBLAzer™ D1-CRE-bla CHO-K1 DA cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
Agonists and antagonists of dopamine D1 receptors are in various stages of clinical trials for the treatment of neurological disorders like Parkinson's disease, Alzheimer's disease, schizophrenia, and psychosis.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ D1-CRE-bla CHO-K1 cells • GeneBLAzer™ D1-CRE-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ D1-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Detection Method | Fluorescence |
| For Use With (Application) | FRET |
| Gene Symbol | DRD1 |
| Product Line | GeneBLAzer |
| Quantity | 2 x 106 Cells |
| Reporter Gene | BLA (Beta-Lactamase) |
| Target Entry | DRD1, D1 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.