Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK 293T Cells

Catalog No. K1713
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 106 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1713 2 x 106 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1713 Supplier Thermo Scientific™ Supplier No. K1713
Only null left
Add to Cart
Add to Cart
Request a Quote

GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK293T cells contain the human Endothelial Differentiation Receptor 3 (EDG3) stably integrated into the GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK293T cell line.

GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK293T cells contain the human Endothelial Differentiation Receptor 3 (EDG3) stably integrated into the GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK293T cell line. GeneBLAzer™ Gα15-NFAT-bla HEK293T cells (Cat. No. K1539) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Promiscuous G protein, Gα15.

The GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK293T cells are functionally validated for Z' and EC50 concentrations of S1P. In addition, GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK293T cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

EDG-3/S1P-3(Sphingosine-1-Phosphate-3) is a Gq/Gi/Go/Gα13/Gα12 coupled GPCR. EDG-3 has been shown to be responsible for bradycardia that has been induced in clinical trials as a side effect of the immunosuppressive drug FTY720 that targets EDG-1. EDG-3 has also been shown to induce vasodilation in response to sphingosylphosphorylcholine (SPC), sphingosine-1-phosphate (S1P), and lysosulfatide (LSF). The EDG-3 Receptor has also been shown to play a cooperative or redundant role in angiogenesis with EDG-1.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_005226.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK293T cells
• GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK293T Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK293T cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol EDG3
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry EDG3
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.