Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ ER Beta DA Assay Kit

Catalog No. K1399
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1399 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1399 Supplier Thermo Scientific™ Supplier No. K1399
Only null left
Add to Cart
Add to Cart

GeneBLAzer™ER beta DA(Division Arrested) cells and ER beta-UAS-bla GripTite™ 293 cells contain the ligand-binding domain (LBD) of the human Estrogen receptor beta (ER beta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla GripTite™ 293 cell line.

GeneBLAzer™UAS-bla GripTite™ 293 cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-ER beta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both ER beta DA cells and ER beta-UAS-bla GripTite™ 293 cells are functionally validated for Z' and EC50 concentrations of 17-beta Estradiol. In addition, ER beta-UAS-bla GripTite™ 293 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time. Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY

Specifications

Alternate Agonists Cholic Acid, Deoxycholic Acid
Antagonists (z)-guggulsterone
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage Shipping: Dry IceStorage: -80°C/Liquid NitrogenSufficient division-arrested cells and substrate to assay one 384-well plate. Includes:ER beta HEK 293 T DA cells (K1399A)LiveBLAzer™-FRET B/G Loading Kit, 70 μg
Detection Method Fluorescence
Gene ID (Entrez) 2100
Gene Symbol ERβ
Primary Agonist Chenodeoxycholic Acid (CDCA)
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry ER beta, ESR2, ERB, NR3A2
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target Nuclear Receptors
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.