Learn More
Thermo Scientific™ GeneBLAzer™ Gα15-NFAT CHO-K1 DA Assay Kit
Description
The GeneBLAzer™ Gα15-NFAT CHO-K1 DA (Division Arrested) cell line contains the promiscuous G protein, Gα15, stably integrated into the CellSensor™ NFAT-bla CHO-K1 (Cat. No. K1078) cell line. NFAT-bla CHO-K1 cells contain a beta-lactamase reporter gene under control of an NFAT response element stably integrated into CHO-K1 cells. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
This product can serve as a negative control in screening assays performed with specific G-protein coupled receptor (GPCR) DA cells that were made from this same background. These cells can also be used in other assays where a sensitive readout to intracellular changes in calcium is needed.
Specifications
Specifications
| Cell State | Division-Arrested Cells |
| Content And Storage | Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol). Includes: • GeneBLAzer™ Gα15-NFAT CHO-K1 DA cells • LiveBLAzer™-FRET B/G Loading Kit, 70 μg • Solution D • GeneBLAzer™ Gα15-NFAT CHO-K1 Assay Protocol • LiveBLAzer™-FRET B/G Loading Protocol • Certificate of Analysis The GeneBLAzer™ Gα15-NFAT CHO-K1 DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Detection Method | Fluorescence |
| Gene Symbol | Gα15 |
| Product Line | GeneBLAzer |
| Quantity | 1 Plate |
| Reporter Gene | BLA (Beta-Lactamase) |
| Cell Line | CHO-K1 |
| System Type | GeneBLAzer™ |
| Druggable Target | GPCR |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.