Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ M1-NFAT-bla Jurkat Cells

Catalog No. K1710
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 106 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1710 5 x 106 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1710 Supplier Thermo Scientific™ Supplier No. K1710
Only null left
Add to Cart
Add to Cart
Request a Quote

GeneBLAzer™ M1-NFAT-bla Jurkat cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla Jurkat cell line.

GeneBLAzer™ M1-NFAT-bla Jurkat cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla Jurkat cell line. CellSensor™ NFAT-bla Jurkat cells (Cat. No. K1077) contain a beta-lactamase (bla) reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.

M1-NFAT-bla Jurkat cells are functionally validated for Z'-factor and EC50 concentrations of carbachol. In addition, M1-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

In the brain, M1 activation mediates 'slow' neuronal excitability. Cortical and hippocampal muscarinic receptors are thought to be important in the attentional aspects of cognition. The predominant receptor subtypes in these brain areas are M1, M3, and M4. Therefore, M1 is a potential target for cognition, Alzheimer's, dementia, and schizophrenia. Studies on knock-out mouse models of M1 are also beginning to reveal potential functions of the receptor. Additional information on the muscarinic receptors can be found in reviews.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000738
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ M1-NFAT-bla Jurkat cells
• GeneBLAzer™ M1-NFAT-bla Jurkat Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ M1-NFAT-bla Jurkat cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol M1
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 5 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry M1
Cell Line Jurkat
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.