Learn More
Invitrogen™ GeneBLAzer™ MC2R-CRE-bla-CHO-K1 Cells
Description
The MC2R-CRE-bla-CHOK1 cells are functionally validated for Z'-factor and EC50 concentrations of ACTH 1-24. In addition, MC2R-CRE-bla-CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
Specifications
Specifications
| Accession Number | NM_000529.2 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: o GeneBLAzer ™ MC2R-CRE-bla-CHO-K1 cells o GeneBLAzer ™ MC2R-CRE -bla CHO-K1 Assay Protocol o Certificate of Analysis The GeneBLAzer™ MC2R-CRE -bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Detection Method | Fluorescence |
| Gene Symbol | MC2R |
| Population Status | Single Cell Clone Origin |
| Product Line | GeneBLAzer |
| Quantity | 2 x 106 Cells |
| Reporter Gene | BLA (Beta-Lactamase) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.