Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ NMUR1-NFAT-bla CHO-K1 Cells

Catalog No. K1719
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 106 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1719 2 x 106 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1719 Supplier Thermo Scientific™ Supplier No. K1719
Only null left
Add to Cart
Add to Cart
Request a Quote

GeneBLAzer™ NMUR1-NFAT-bla CHO-K1 cells contain the human Neuromedin U-25 receptor 1 (NMUR1) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

GeneBLAzer™ NMUR1-NFAT-bla CHO-K1 cells contain the human Neuromedin U-25 receptor 1 (NMUR1) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.

The GeneBLAzer™ NMUR1-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Neuromedin U-25 In addition, GeneBLAzer™ NMUR1-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary. Division Arrested cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.

Two receptors for neuromedin U-25 have been identified through various studies and are 50% homologous to each other. The receptors have different patterns of expression; NMUR1 localizes to the peripheral organs, including the gastrointestinal tract, urogenital system, and kidneys. The central nervous system (CNS) was found to express much lower amounts of NMUR1 than the peripheral organ systems. In contrast, NMUR2 is expressed primarily in the CNS. Both receptors are believed to couple to the Gq family of proteins upon ligand binding. The physiological roles for these receptors are unknown, but some recent animal studies have implicated NMUR1 in inflammation and the regulation of body weight and feeding.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_006056.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ NMUR1-NFAT-bla CHO-K1 cells
• GeneBLAzer™ NMUR1-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ NMUR1-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol NUMR1
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry NMUR1
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.