Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ P2RY6 CHO-K1 DA Assay Kit

Catalog No. K1327
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1327 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1327 Supplier Thermo Scientific™ Supplier No. K1327
Only null left
Add to Cart
Add to Cart

GeneBLAzer™ P2RY6 CHO-K1 DA (Division Arrested) cells and P2RY6-NFAT-bla CHO-K1 cells contain the human purinergic receptor P2, G protein-coupled, 6 (P2RY6) receptor stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

This cell line is available as dividing cells by requesting a quote for K1253 from dividingcells@invitrogen.com

GeneBLAzer™ P2RY6 CHO-K1 DA (Division Arrested) cells and P2RY6-NFAT-bla CHO-K1 cells contain the human purinergic receptor P2, G protein-coupled, 6 (P2RY6) receptor stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both P2RY6 CHO-K1 DA cells and P2RY6-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Uridine 5'-diphosphate (UDP); (Figure 1). In addition, P2RY6-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

P2RY6 has been found in a variety of rat tissue such as; placenta, spleen, thymus, lung, stomach, intestine, mesentery, heart, and aorta. P2RY6 is activated by UDP, which functions as a full agonist. P2RY6 is activated either weakly or not at all by additional extracellular nucleotides such as UTP, ATP, and ADP. P2RY6 has been implicated to be involved in ion transport, vasodilatation, and contraction of smooth muscle.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_004154.3
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage GeneBLAzer™ P2RY6 CHO-K1 DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
• P2RY6 CHO-K1 DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D, 1 mL
• 2 protocols
• Certificate of Analysis

GeneBLAzer™ P2RY6 CHO-K1 DA cells
Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• P2RY6 CHO-K1 DA cells
• 1 protocol
• Certificate of Analysis

GeneBLAzer™ P2RY6-NFAT-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• P2RY6-NFAT-bla CHO-K1 cells
• 1 protocol
• Certificate of Analysis

Storage:
GeneBLAzer™ P2RY6 CHO-K1 DA Cells and P2RY6-NFAT-bla CHO-K1 Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.
Detection Method Fluorescence
Gene ID (Entrez) 5031
Gene Symbol P2RY6
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry P2RY6
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.