Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ PPAR alpha UAS-bla HEK 293T Cellular Assay Kit

Catalog No. K1875
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1875 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1875 Supplier Thermo Scientific™ Supplier No. K1875
Only null left
Add to Cart
Add to Cart

The GeneBLAzer™ PPAR alpha-UAS-bla HEK 293T Cellular Assay provides an accurate, sensitive, and easy-to-use method for monitoring the cellular response of PPAR alpha to drug candidates or other stimuli.

The GeneBLAzer™ PPAR alpha-UAS-bla HEK 293T Cellular Assay provides an accurate, sensitive, and easy-to-use method for monitoring the cellular response of PPAR alpha to drug candidates or other stimuli.

Features of the The GeneBLAzer™ PPAR alpha-UAS-bla HEK 293T Cellular Assay:

  • Convenience—plate cells and perform assay without additional cell culture steps
  • Low Noise—the kit uses a Fluorescence Resonance Energy Transfer (FRET)-based ratiometric readout for minimum noise and error
  • Low Background—the GAL4 fusion prevents any background from endogenous receptor

The Cells

The GeneBLAzer™ PPAR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human peroxisome proliferator-activated receptor alpha (PPAR alpha) fused to the DNA-binding domain of GAL4 transiently transduced (via BacMam virus) into the GeneBLAzer™ UAS-bla HEK293T cell line (Cat. No. K1711). GeneBLAzer™ UAS-bla HEK 293T cells stably express a β-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS).

The Assay

When an agonist binds to the LBD of the GAL4 (DBD)-PPAR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of β-lactamase. The FRET ratiometric readout from the live-cell substrate reduces the absolute and relative errors that can mask the underlying biological response of interest. PPAR alpha UAS-bla HEK 293T cells have been tested for assay performance using variable assay conditions, including cell number, stimulation time, substrate loading time and have been validated for Z' and EC50 concentrations of GW7647. Additional testing data using alternate stimuli are also available. See the 'validation packet' in the documents below to view the data.

Speed and Convenience

Because PPAR alpha UAS-bla HEK 293T cells are transiently transduced with BacMam virus, they are ready to be used in an assay without any additional cell culture steps. Just plate the cells and assay as outlined in the protocol.

Please contact discoveryservices@lifetech.com to inquire about obtaining these cells at economical bulk scales.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_005036
Assay Entry Cell-based beta lactamase reporter gene
Cell State Transiently Transduced
Content And Storage • PPAR alpha UAS-bla HEK293T cells (K1875A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol PPARA
Human Protein Class NHR
Product Line GeneBLAzer
Quantity 1 Plate
Readout End Point
Reporter Gene BLA (Beta-Lactamase)
Target Entry PPARA, PPAR alpha, NR1C1
Type Cellular Assay Kit
Cell Line HEK 293T
System Type GeneBLAzer™
Druggable Target Nuclear Receptors
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.