Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ TACR2-NFAT-bla CHO-K1 Cells

Catalog No. K1844
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 x 106 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1844 8 x 106 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1844 Supplier Thermo Scientific™ Supplier No. K1844
Only null left
Add to Cart
Add to Cart
Request a Quote

GeneBLAzer™ TACR2-NFAT-bla CHO-K1 cells contain the human Tachykinin receptor 2 (TACR2), (Accession No. NM_001057) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

GeneBLAzer™ TACR2-NFAT-bla CHO-K1 cells contain the human Tachykinin receptor 2 (TACR2), (Accession No. NM_001057) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the NFAT Response Element. TACR2-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of (Nle10)-Neurokinin A (4-10) (Figure 1). In addition, TACR2-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_001057
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ TACR2-NFAT-bla CHO-K1 cells
• GeneBLAzer™ TACR2-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ TACR2-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol TACR2
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Pool of Stable Cells
Product Line GeneBLAzer
Quantity 8 x 106 Cells
Readout End Point
Target Entry TACR2
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.