Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ TRα-UAS-bla HEK 293T Assay Kit

Catalog No. K1684
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1684 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1684 Supplier Thermo Scientific™ Supplier No. K1684

Please call customer service for current pricing. 1-800-766-7000

TR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Thyroid hormone receptor alpha(TR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line.

TR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Thyroid hormone receptor alpha(TR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-TR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. TR alpha-UASbla HEK 293T 293 cells are functionally validated for Z' and EC50 concentrations of T3 Thryoid hormone. In addition, TR alpha-UAS-bla HEK 293T 293 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY

Specifications

Alternate Agonists 9-cis-retinoic acid, TTNPB, CD437, AM 580
Antagonists AGN 193109
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage TR alpha-UAS-bla HEK 293T cells (K1260)

Shipping: Dry Ice
Storage:-80°C (liquid nitrogen)
Detection Method Fluorescence
Primary Agonist All-trans-retinoic Acid
Product Line GeneBLAzer
Quantity 1 Plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry THRA, TR alpha, NR1A1
Type Detection Assay Kit
Cell Line HEK 293T
System Type GeneBLAzer™
Druggable Target Nuclear Receptors
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.