Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ VDR DA Assay Kit

Catalog No. K1417
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1417 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1417 Supplier Thermo Scientific™ Supplier No. K1417
Only null left
Add to Cart
Add to Cart

GeneBLAzer™ VDR DA (Division Arrested) cells and VDR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Vitamin D receptor (VDR) fused to the DNAbinding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line.

GeneBLAzer™ VDR DA (Division Arrested) cells and VDR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Vitamin D receptor (VDR) fused to the DNAbinding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™ UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-VDR (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both VDR DA cells and VDR-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of Calcitriol (1α, 25-dihydroxyvitamin D3). In addition, VDR-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• VDR HEK 293T DA cells (K1417A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Detection Method Fluorescence
Gene ID (Entrez) 7421
Gene Symbol VDR
Primary Agonist TO901317
Product Line GeneBLAzer
Quantity 1 Plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry VDR, NR1I1
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target Nuclear Receptors
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.