Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GenePrint 10 System, 50 reactions

Catalog No. PRB9510 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
GenePrint 10 System, 50 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRB9510 GenePrint 10 System, 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRB9510 Supplier Promega Supplier No. B9510
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The GenePrint 10 System allows co-amplification and three-color detection of nine human loci, including the ASN-0002 loci (TH01, TPOX, vWA, Amelogenin, CSF1PO, D16S539, D7S820, D13S317 and D5S818) as well as D21S11.

  • STR Analysis for Human Cell Line Authentication
  • Amplifies ANSI−0002−Recommended Loci
  • Tolerates higher DNA amounts. Better balance for aneuploid samples
  • Reduced PCR Time. Amplification in less than 1.5 hours

Specifications

Content And Storage -30C TO -10C
Control Template 2800M Control DNA (positive control)
Form Liquid
Format Kit
PCR Method Standard PCR
Polymerase Hot-start Thermostable DNA Polymerase
Technique STR (Short Tandem Repeat) Analysis
For Use With (Equipment) Capillary Electrophoresis System, Genetic Analyzer, Thermal Cycler
Genotyping Target Short Tandem Repeats
Includes Label or Dye Yes
Kit Type STR Analysis Detection Kit
Matrix and Size Standards Internal Lane Standard 600
No. of Tests 50 Test
Primer STR Primer Mix
Product Line GenePrint
Product Type STR Analysis Kit
Quantity 50 reactions
Research Category Cell Line Authentication
Shipping Condition Dry Ice
Sufficient For 50 Reactions
Target Loci TH01, TPOX, vWA, Amelogenin, CSF1PO, D16S539, D7S820, D13S317, D5S818, D21S11
Target Organism Class Human
For Use With (Application) STR-based Human Identification
Compatibility Capillary Electrophoresis System, Genetic Analyzer, Thermal Cycler
Final Product Type Amplified DNA
Hot Start Built-In Hot Start
No. of Reactions 50 Reactions
Overhang 3'-A
Reaction Format Kit
Reagent Type STR Analysis Reagent
Sample Type Human Genomic DNA
Starting Material 10 ng DNA
Volume 25 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.