Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GenePrint 5C Matrix Standard, 5 preps

Catalog No. PRB1930 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
GenePrint 5C Matrix Standard, 5 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRB1930 GenePrint 5C Matrix Standard, 5 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRB1930 Supplier Promega Supplier No. B1930
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The GenePrint 5C Matrix Standard allows the GenePrint 24 System to be analyzed on the Applied Biosystems 3130, 3130xl, 3500 and 3500xL Genetic Analyzers.

  • Matrix Fragments for use with the GenePrint(R) 24 System
  • Labeled with five fluorescent dyes: Fluorescein, JOE, TMR−ET, CXR−ET and WEN
  • Used for spectral calibration on Applied Biosystems(R) 3130, 3130xl, 3500 and 3500xL Genetic Analyzers

Specifications

Calibration Type Spectral Calibration
Content And Storage -30C TO +10C
Format Kit
Technique Spectral Calibration
For Use With (Equipment) Capillary Electrophoresis System, Genetic Analyzer
Includes Label or Dye Yes
Matrix and Size Standards GenePrint™ 5C Matrix Standard
No. of Tests 5 Test
Product Line GenePrint
Product Type Calibration Standard
Quantity 5 preps
Research Category Forensics
Shipping Condition Dry Ice
Sufficient For 5 Preps
Compatibility Capillary Electrophoresis System, Genetic Analyzer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.