Learn More
Surgical Design™ General Purpose Lab Spatula
Catalog No. p-7162735 Shop All Fisherbrand Products
Click to view available options
Length (English) Blade:
4 in.
6 in.
Fisherbrand™ General Purpose Lab Spatula is used to transfer sticky or corrosive materials without scratching or damaging container surfaces
  • Non-stick coating
  • Multiple sizes allow for varied laboratory applications.
Specifications
Quantity 1
Coating PTFE
Product Type General Purpose Spatula
For Use With (Application) Non-stick lab applications
Length (English) Blade 6 in.
Handle Material Wooden Handle
Material Stainless Steel
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.