Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Genetics I Kit
SCI_ED

Catalog No. s30806 Shop All AquaPhoenix Scientific Products
Click to view available options
Class Size:
24 Students

Kemtec™ Genetics I Kit includes three activities that introduce students to Punnet Squares, and other genetic topics

  • Make genetics personal by helping students assess their own genetic traits
  • Model homologous chromosomes to demonstrate how reproduction results in the inheritance of genetic variation
  • Use Punnett squares to predict the distribution and variation of expressed traits in humans, plants, and animals
  • Introduces allele frequency, phenotypes, genotypes, monohybrid and dihybrid crosses, test crosses, dominance, incomplete dominance, codominance, and recessives
  • Addresses National Standards for Grade Levels 6-10
  • MS-LS3-2, MS-LS3.A, MS-LS3.B, HS-LS3-3

Specifications

Certifications/Compliance MS-LS3-2, MS-LS3.A, MS-LS3.B, HS-LS3-3
Product Type Genetics
Grade 6 to 10
Age 11 to 16
For Use With (Application) Grades 6-10
Includes Instructor's Manual with Reproducible Experiments and Worksheets, PTC Paper, Colored Plastic Marker Chips, Homologus Chromosome Sticks
Weight (English) 4 lb.
Dimensions (L x W x H) 13 x 12 x 5 in.
Class Size 24 Students
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.