Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Genistein

Click to view available options
Quantity:
10 mg
50 mg
Description
Genistein is a phytoestrogen with a wide range of biological actions. Genistein inhibits protein tyrosine kinases including epidermal growth factor receptor kinase, binds to PPARγ and estrogen receptors and acts as an agonist at GPER. Genistein inhibits aggregation and fibrilization of Aβ42 and amylin37 (hiAPP). In Huntington's disease fibroblast cells, Genistein reduces intracellular mutant huntingtin (mHTT) aggregates and induces degradation of mHTT; in a cellular model of Huntington's disease, Genistein corrects the mutant phenotype through stimulation of autophagy.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 270.24 |
| Purity | >98% |
| Molecular Weight (g/mol) | 270.24 |
| Chemical Name or Material | Genistein |
Safety and Handling
| Recommended Storage | Desiccate at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction