Learn More
Amsbio LLC (formerly Genlantis) TurboCells™ BL21Competent Cells
                    Click to view available options
                                Product Type:
                                BL21 (DE3) Competent E. coli
                    Description
- Achieve high-level recombinant protein expression with the T7 expression system
- No hour-long recovery step or inconvenient 42°C water bath
- Convenient and efficient single-use packaging
- Two useful genotypes
Specifications
Specifications
| For Use With (Application) | High-level E. coli expression using 3 min. transformation protocol | 
| Product Type | BL21 (DE3) Competent E. coli | 
| Quantity | 20 x 50 μL | 