Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gentamicin Monoclonal Antibody (GB26.16)
GREENER_CHOICE

Catalog No. 01671956
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-956 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-956 Supplier Invitrogen™ Supplier No. MA110726
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10726 detects Gentamicin. MA1-10726 has been successfully used in ELISA applications. The MA1-10726 immunogen is Gentamicin, conjugated to KLH.

Gentamicin is an aminoglycoside antibiotic, used to treat many types of bacterial infections, particularly those caused by Gram-negative bacteria. However, gentamicin is not used for Neisseria gonorrhoeae, Neisseria meningitidis or Legionella pneumophila bacterial infections (because of the risk of the patient going into shock from lipid A endotoxin found in certain gram negative organisms).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gentamicin
Applications ELISA
Classification Monoclonal
Clone GB26.16
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Gentacycol; Garamycin; Gentavet; Gentamicinum; C21H43N5O7
Host Species Mouse
Immunogen Gentamicin, conjugated to KLH.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.