Learn More
Description
- Smooth, uniform surface to support seed placement and growth
- High water absorption and retention capacity for consistent moisture
- Free from toxins and contaminants that may affect germination
- Widely used in agricultural labs, seed companies, and research institutes
- Plant growth studies
- Agricultural and botanical laboratories
Specifications
Specifications
| Color | Yellow |
| Format | Roll |
| For Use With (Equipment) | Conical Funnel |
| Material | Cellulose |
| Type | Qualitative Filter Paper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.