Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Antibody (012), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB354583 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354583 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354583 Supplier Novus Biologicals Supplier No. NBP289203JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen GFAP
Applications Western Blot
Classification Monoclonal
Clone 012
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human GFAP.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2670
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.