Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Antibody (ASTRO/1974R), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB380595 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380595 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380595 Supplier Novus Biologicals Supplier No. NBP308431R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen GFAP
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ASTRO/1974R
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
Host Species Rabbit
Immunogen Recombinant human full-length GFAP protein (Uniprot: P14136)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2670
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.