Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Antibody, Novus Biologicals™
SDP

Catalog No. NB018354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018354 0.025 mL
NBP105198 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018354 Supplier Novus Biologicals Supplier No. NBP1051980.025ML
Add to Cart
Add to Cart

Chicken Polyclonal Antibody has been used in 19 publications

GFAP Polyclonal specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.

Specifications

Antigen GFAP
Applications Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:1000-1:5000, Immunohistochemistry-Frozen 1:1000-1:5000, Immunohistochemistry Free-Floating 1:1000-1:5000
Formulation PBS with 5mM Sodium Azide
Gene Accession No. Q28115
Gene Alias FLJ45472, GFAP astrocytes, glial fibrillary acidic protein
Gene Symbols GFAP
Host Species Chicken
Immunogen Recombinant full length human GFAP isotype 1 expressed in and purified from E. coli.
Molecular Weight of Antigen 55 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Astrocyte Markers, Cancer, Cytoskeleton Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2670
Test Specificity Human, mouse, rat, chicken, bovine and feline. Predicted to work with all mammals.
Target Species Bovine, Rat, Pig, Equine, Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.