Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFAP Antibody, Novus Biologicals™

Chicken Polyclonal Antibody has been used in 19 publications
$214.50 - $518.50
Specifications
Antigen | GFAP |
---|---|
Applications | Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Precipitation |
Description
GFAP Polyclonal specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.Specifications
GFAP | |
Polyclonal | |
Precipitation | |
RUO | |
Q28115 | |
2670 | |
IgY | |
Ammonium sulfate precipitation | |
Human, mouse, rat, chicken, bovine and feline. Predicted to work with all mammals. |
Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Chicken | |
Bovine, Rat, Pig, Equine, Human, Mouse | |
FLJ45472, GFAP astrocytes, glial fibrillary acidic protein | |
GFAP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title