Learn More
GFAP (Glial Fibrillary Acidic Protein) Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: ASTRO6
Isotype: IgG1
Species Reactivity: Human, Pig, Rat and Chicken. Others not known.
Epitope: Not determined
Immunogen: Glial Fibrillary Acidic Protein
Molecular Weight: 51-52 kDa
Positive Control: IMR-5 cells, Brain or astrocytoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Specifications
| Antigen | GFAP (Glial Fibrillary Acidic Protein) |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | ASTRO6 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Glial Fibrillary Acidic Protein |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.