Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Rabbit anti-Human, Clone: 12, Novus Biologicals™
SDP

Catalog No. NB019890 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019890 100 μg
NB019891 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019890 Supplier Novus Biologicals Supplier No. NBP289203100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody has been used in 2 publications

GFAP Monoclonal specifically detects GFAP in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen GFAP
Applications Western Blot
Classification Monoclonal
Clone 12
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation 0.2 um filtered solution in PBS
Gene Alias FLJ45472, GFAP astrocytes, glial fibrillary acidic protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the human GFAP
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Neuroscience, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2670
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.