Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFER/ALR Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB041657 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB041657 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB041657 Supplier Novus Biologicals Supplier No. NBP297019AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GFER/ALR Polyclonal specifically detects GFER/ALR in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GFER/ALR
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ALRFAD-linked sulfhydryl oxidase ALR, Augmenter of liver regeneration, EC 1.8.3.2, ERV1 homolog, ERV1hepatopoietin protein, erv1-like growth factor, growth factor, augmenter of liver regeneration, growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration), Hepatopoietin, hERV1, HERV1hepatic regenerative stimulation substance, HPO, HPO1, HPO2, HSS, truncated augmenter of liver regeneration
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant GFER/ALR (NP_005253.3; Met1-Asp125).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2671
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.