Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFI-1 Antibody (OTI6E8), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP270827FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270827FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270827FR Supplier Novus Biologicals Supplier No. NBP270827FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GFI-1 Monoclonal specifically detects GFI-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen GFI-1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6E8
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias GFI-1, growth factor independence-1, growth factor independent 1, growth factor independent 1 transcription repressor, Growth factor independent protein 1, SCN2, Zinc finger protein 163, zinc finger protein Gfi-1, ZNF163FLJ94509
Gene Symbols GFI1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-180 of human GFI-1 (NP_005254) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2672.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.