Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFOD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157621
Description
GFOD1 Polyclonal specifically detects GFOD1 in Human samples. It is validated for Western Blot.Specifications
| GFOD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ADG-90, chromosome 6 open reading frame 114, FLJ20330, hypothetical protein LOC85411, Protein ADG-90, RP11-501I19.1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Chicken: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Guinea pig: 85%; Rabbit: 85%; Zebrafish: 84%. | |
| Human, Mouse, Rat, Pig, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NXC2 | |
| GFOD1 | |
| Synthetic peptides corresponding to GFOD1(glucose-fructose oxidoreductase domain containing 1) The peptide sequence was selected from the middle region of GFOD1. Peptide sequence NSLLPEKAFSDIPSPYLRGTIKMMQAVRQAFQDQDDRRTWDGRPLTMAAT. | |
| 100 μL | |
| Lipid and Metabolism | |
| 54438 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction