Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody (208), PerCP, Novus Biologicals™
SDP

Catalog No. NB316645 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316645 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316645 Supplier Novus Biologicals Supplier No. NBP290178PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GFP Monoclonal antibody specifically detects GFP in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA

Specifications

Antigen GFP
Applications Western Blot, ELISA, Immunoprecipitation, ELISA
Classification Monoclonal
Clone 208
Conjugate PerCP
Dilution Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA Detection
Formulation PBS
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Aequorea victoria GFP (Accession#: AAB65663; Ser2-Lys238).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.