Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody (454505R), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB370520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370520 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370520 Supplier Novus Biologicals Supplier No. FAB42401RM
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GFP Monoclonal specifically detects GFP in Multi-species samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen GFP
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 454505R
Conjugate DyLight 594
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Host Species Mouse
Immunogen E.coli-derived recombinant GFPuv, Ser2-Lys238, Accession # P42212
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.