Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody (4B10B2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222111 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222111 0.1 mL
NBP222111SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222111 Supplier Novus Biologicals Supplier No. NBP222111
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GFP
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4B10B2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000. Use reported by customer review, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Host Species Mouse
Immunogen Purified recombinant fragment of GFP expressed in E. coli.
Molecular Weight of Antigen 27 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers
Primary or Secondary Primary
Test Specificity Non-species specific. Reacts with GFP.
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.