Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody (4B10B2), PE, Novus Biologicals™
SDP

Catalog No. NBP222111PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222111PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222111PE Supplier Novus Biologicals Supplier No. NBP222111PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Flow Cytometry.

Specifications

Antigen GFP
Applications Flow Cytometry
Classification Monoclonal
Clone 4B10B2
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Host Species Mouse
Immunogen Purified recombinant fragment of GFP expressed in E. coli.
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.