Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody (9F9.F9), Novus Biologicals™
SDP

Catalog No. NB600597SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600597SS 0.025 mg
NB600597 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600597SS Supplier Novus Biologicals Supplier No. NB600597SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 44 publications

GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, In vitro assay, Dot Blot, Knockdown Validated.

Specifications

Antigen GFP
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, In vitro Assay, Dot Blot, KnockDown
Classification Monoclonal
Clone 9F9.F9
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:3000-1:30000, Flow Cytometry 1:1:10 - 1:1000, ELISA 1:10000-1:30000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunoblotting, In vitro assay, Dot Blot 1:100 - 1:2000, Knockdown Validated
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Alias clone 9F9.F9, Enhanced Green Fluorescent Protein, GFP monoclonal, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Mouse
Immunogen Recombinant Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246 aa) derived from the jellyfish Aequorea victoria.
Molecular Weight of Antigen 27 kDa
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers
Primary or Secondary Primary
Test Specificity Reactivity is observed against recombinant Green Fluorescent Protein from Aequorea victoria by both Western blot and ELISA. No reaction is seen against RFP.
Target Species Jellyfish
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.