Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody (9F9.F9), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 44 publications
$207.00 - $512.00
Specifications
Antigen | GFP |
---|---|
Clone | 9F9.F9 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:3000-1:30000, Flow Cytometry 1:1:10 - 1:1000, ELISA 1:10000-1:30000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunoblotting, In vitro assay, Dot Blot 1:100 - 1:2000, Knockdown Validated |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, In vitro Assay, Dot Blot, KnockDown |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB600597SS
![]() |
Novus Biologicals
NB600597SS |
0.025 mg |
Each for $207.00
|
|
|||||
NB600597
![]() |
Novus Biologicals
NB600597 |
0.1 mg |
Each for $512.00
|
|
|||||
Description
GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, In vitro assay, Dot Blot, Knockdown Validated.Specifications
GFP | |
1 mg/ml | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, In vitro Assay, Dot Blot, KnockDown | |
Unconjugated | |
Mouse | |
Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
clone 9F9.F9, Enhanced Green Fluorescent Protein, GFP monoclonal, Green Fluorescent Protein, green fluorescent protein (gfp) | |
Recombinant Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246 aa) derived from the jellyfish Aequorea victoria. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. | |
27 kDa |
9F9.F9 | |
Western Blot 1:3000-1:30000, Flow Cytometry 1:1:10 - 1:1000, ELISA 1:10000-1:30000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunoblotting, In vitro assay, Dot Blot 1:100 - 1:2000, Knockdown Validated | |
Monoclonal | |
Purified | |
RUO | |
Jellyfish | |
GFP | |
IgG1 κ | |
Protein A purified | |
Reactivity is observed against recombinant Green Fluorescent Protein from Aequorea victoria by both Western blot and ELISA. No reaction is seen against RFP. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title