Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB272115 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272115 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272115 Supplier Novus Biologicals Supplier No. NBP250059AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GFP Polyclonal antibody specifically detects GFP in Nonspecies specific samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen GFP
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Host Species Rabbit
Immunogen Recombinant AcGFP expressed in and purified from E. coli. (UniProt: Q6YGZ0)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.