Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB1001771 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001771 0.1 mL
NB1001771SS 0.02 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001771 Supplier Novus Biologicals Supplier No. NB1001771
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 9 publications

GFP Polyclonal specifically detects GFP in Mouse, Non-species specific samples. It is validated for Dot Blot, Flow Cytometry, Fluorophore-linked immunosorbent assay, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), KnockDown, Proximity Ligation Assay, Western Blot.

Specifications

Antigen GFP
Applications Dot Blot, Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), KnockDown, Proximity Ligation Assay, Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate FITC
Dilution Western Blot >1:10000, Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:2500, Immunohistochemistry-Frozen, Proximity Ligation Assay, Dot Blot, Fluorophore-linked immunosorbent assay >1:20000, Knockout Validated, Knockdown Validated
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Goat
Immunogen The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. (Uniprot: P42212)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity No reaction was observed against Human, Mouse and Rat Serum Proteins.
Target Species Mouse, Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.