Learn More
Description
Specifications
Specifications
| Antigen | GFP |
| Applications | Dot Blot, Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), KnockDown, Proximity Ligation Assay, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0mg/mL |
| Conjugate | FITC |
| Dilution | Western Blot >1:10000, Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:2500, Immunohistochemistry-Frozen, Proximity Ligation Assay, Dot Blot, Fluorophore-linked immunosorbent assay >1:20000, Knockout Validated, Knockdown Validated |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide |
| Gene Alias | Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) |
| Gene Symbols | GFP |
| Host Species | Goat |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
