Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB1001183 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001183 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001183 Supplier Novus Biologicals Supplier No. NB1001183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen GFP
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate FITC
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:2000-1:10000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:2500, Immunohistochemistry-Frozen
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Accession No. P42212
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Rabbit
Immunogen The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria.
Molecular Weight of Antigen 27 kDa
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity GFP from Aequorea victoria
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.