Learn More
Description
Specifications
Specifications
| Antigen | GFP |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, Immunohistochemistry (Free Floating), Immunohistochemistry, KnockDown |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockdown Validated |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) |
| Host Species | Rabbit |
| Immunogen | The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
