Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, Novus Biologicals™
SDP

Catalog No. NB10056401T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056401T 0.025 mg
NB10056401 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056401T Supplier Novus Biologicals Supplier No. NB10056401SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot.

Specifications

Antigen GFP
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 102-118 (KDDGNYKTRAEVKFEGD) of GFP were used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers
Primary or Secondary Primary
Test Specificity The GFP amino acid sequence used as immunogen for this antibody is 100% homologous in YFP (Yellow) and CFP (Cyan).
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.