Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, Novus Biologicals™
SDP

Catalog No. NB600308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600308 0.1 mg
NB600308SS 0.025 mg
NB456507 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB600308 Supplier Novus Biologicals Supplier No. NB600308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Electron Microscopy, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockdown Validated.

Specifications

Antigen GFP
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Immunohistochemistry (Free Floating), Immunohistochemistry, KnockDown
Classification Polyclonal
Concentration 0.99 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Flow Cytometry 1:10-1:1000, ELISA 1:20000-1:120000, Immunohistochemistry 1:200-1:3000, Immunocytochemistry/Immunofluorescence 1:500-1:5000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:250, Dot Blot, Electron Microscopy 1:10-1:500, Immunohistochemistry Free-Floating, Immunohistochemistry, KnockDown Validated
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P42212
Gene Alias anti-gfp (green fluorescent protein) pab, Enhanced Green Fluorescent Protein, GFP immunofluorescence, GFP immunoprecipitation, GFP mouse, GFP staining, GFP western blot, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Rabbit
Immunogen The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria.
Molecular Weight of Antigen 27 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers
Primary or Secondary Primary
Test Specificity No reaction was observed against Human, Mouse or Rat serum proteins.
Target Species Non-species specific, C. elegans, Plant
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.