Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $426.00
Specifications
Antigen | GFP |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056401T
![]() |
Novus Biologicals
NB10056401SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056401
![]() |
Novus Biologicals
NB10056401 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot.Specifications
GFP | |
Western Blot 0.5-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Non-species specific | |
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
IgG | |
Protein G purified | |
The GFP amino acid sequence used as immunogen for this antibody is 100% homologous in YFP (Yellow) and CFP (Cyan). |
1.0 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
PBS with 0.05% Sodium Azide | |
A synthetic peptide corresponding to amino acids 102-118 (KDDGNYKTRAEVKFEGD) of GFP were used as immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title