Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, Novus Biologicals™
SDP

Catalog No. NB017500 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017500 0.1 mL
NB10062622 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017500 Supplier Novus Biologicals Supplier No. NB100626220.1ML
Add to Cart
Add to Cart

Sheep Polyclonal Antibody has been used in 4 publications

GFP Polyclonal specifically detects GFP in Drosophila, Human, Mouse, Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen GFP
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000, Immunohistochemistry-Paraffin
Formulation PBS with 0.09% Sodium Azide
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Sheep
Immunogen Green fluorescent protein from jellyfish
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers
Primary or Secondary Primary
Test Specificity NB100-62622 recognizes green fluorescent protein (GFP), a 27 kDa protein which is derived from the jellyfish Aequorea victoria. GFP fluoresces green (509 nm) when excited by blue light (395 nm) and is commonly used as a marker of gene expression.
Target Species Drosophila, Human, Mouse, Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.