Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, Novus Biologicals™
SDP

Sheep Polyclonal Antibody has been used in 4 publications

$206.50 - $487.50

Specifications

Antigen GFP
Concentration 5.0 mg/mL
Dilution ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000, Immunohistochemistry-Paraffin
Applications Immunohistochemistry (Paraffin), Immunofluorescence, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017500
SDP
View Documents
Novus Biologicals
NB100626220.1ML
0.1 mL
Each for $206.50
Only null left
Add to Cart
 
NB10062622
SDP
View Documents
Novus Biologicals
NB10062622
0.5 mL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

GFP
ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Jellyfish
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Green fluorescent protein from jellyfish
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
5.0 mg/mL
Immunohistochemistry (Paraffin), Immunofluorescence, ELISA, Immunohistochemistry, Immunocytochemistry
Unconjugated
Sheep
Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers
PBS with 0.09% Sodium Azide
GFP
IgG
Protein G purified
NB100-62622 recognizes green fluorescent protein (GFP), a 27 kDa protein which is derived from the jellyfish Aequorea victoria. GFP fluoresces green (509 nm) when excited by blue light (395 nm) and is commonly used as a marker of gene expression.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.