Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, Novus Biologicals™
SDP

Catalog No. NB1001614 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001614 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001614 Supplier Novus Biologicals Supplier No. NB1001614
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

GFP Polyclonal specifically detects GFP in Chicken, Drosophila, Firefly, Human, Jellyfish, Mouse, Non-species specific, Primate, Rat, Yeast, Zebrafish samples. It is validated for ChIP Assay, ELISA, Flow Cytometry, IF/IHC, IHC/IF, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vivo Assay, Western Blot.

Specifications

Antigen GFP
Applications ChIP Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vivo Assay
Classification Polyclonal
Concentration 10.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:10000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:2000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, In vivo assay, Immunohistochemistry Free-Floating
Formulation 10 mM PBS (0.9% isotonic, w/v, pH 7.2) and 50% (v/v) Glycerol with 0.02% Sodium Azide
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Chicken
Immunogen Chickens were immunized with purified recombinant green fluorescent protein (GFP) emulsified in Freund's adjuvant.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers
Primary or Secondary Primary
Test Specificity GFP
Target Species Chicken, Drosophila, Drosophila, Firefly, Human, Jellyfish, Mouse, Non-species specific, Primate, Rat, Zebrafish
Content And Storage Store at -20C in the dark. Avoid freeze-thaw cycles.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.