Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Antibody, PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N60308PECY5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N60308PECY5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N60308PECY5 Supplier Novus Biologicals Supplier No. NB600308PECY55
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Flow Cytometry, Knockdown Validated.

Specifications

Antigen GFP
Applications Flow Cytometry, KnockDown
Classification Polyclonal
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Rabbit
Immunogen The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers
Primary or Secondary Primary
Test Specificity No reaction was observed against Human, Mouse or Rat serum proteins.
Target Species Non-species specific, C. elegans, Plant
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.