Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $561.50
Specifications
Antigen | GFP |
---|---|
Clone | 454505R |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB42401RSP
![]() |
R&D Systems
MAB42401RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB42401RP
![]() |
R&D Systems
MAB42401R100 |
100 μg |
Each for $561.50
|
|
|||||
Description
GFP Monoclonal specifically detects GFP in Multi-Species samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
GFP | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL | |
Monoclonal | |
Mouse | |
Multi-species | |
eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp) | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
454505R | |
Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
RUO | |
P42212 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Stains eGFP transfectants but not irrelevant transfectants in flow cytometry. Detects recombinant GFPuv in Western blots. For optimal Western blot results, we recommend R&D Systems Goat Anti-GFP Antigen Affinity-purified Polyclonal Antibody (Catalog # AF4240 ) or Novus Biologicals Mouse Anti-GFP Monoclonal Antibody (Catalog # NBP2-2211 ). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title